missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-5-trimethylsilyl-4-pentyne, 97%

Manufacturer:  Alfa Aesar™ H5339306

 View more versions of this product

Catalog No. AAH5339306



Description

Description

Applications
1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1-Chloro-5-trimethylsilyl-4-pentyne
C8H15ClSi
MFCD00671353
UPVHIBLXQNNITH-UHFFFAOYSA-N
5-chloropent-1-ynyl(trimethyl)silane
2757942
97%
5g
77113-48-5
Cl(CH2)3C≡CSi(CH3)3
5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 1-chloro-5-trimethylsilyl-4-pentyne, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, 5-chloro-1-pentynyl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
C[Si](C)(C)C#CCCCCl
174.743
174.75
75°C to 77°C (21mmHg)
65°C (149°F)

Chemical Identifiers

77113-48-5
174.743
UPVHIBLXQNNITH-UHFFFAOYSA-N
2757942
C[Si](C)(C)C#CCCCCl
C8H15ClSi
MFCD00671353
5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 1-chloro-5-trimethylsilyl-4-pentyne, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, 5-chloro-1-pentynyl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
5-chloropent-1-ynyl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents