Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-5-methylhexane, 98%

Chemical Identifiers

Chemical Name or Material 1-Chloro-5-methylhexane
CAS 33240-56-1
Molecular Formula (CH3)2CH(CH2)4Cl
Formula Weight 134.65
MDL Number MFCD00798584
UN Number UN1993
Synonym Isoheptyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2013306

  SDS
alfa aesar™
B2013306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2013314

  SDS
alfa aesar™
B2013314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.65
Quantity 5g
UN Number UN1993
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 38°C (100°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Molecular Formula (CH3)2CH(CH2)4Cl
Assay 98%
MDL Number MFCD00798584
CAS 33240-56-1
Synonym Isoheptyl chloride
Health Hazard 1 Warning
Chemical Name or Material 1-Chloro-5-methylhexane
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.