Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-4-(trifluoromethoxy)benzene, 98%

$171.77 - $171.77

Chemical Identifiers

Chemical Name or Material 1-Chloro-4-(trifluoromethoxy)benzene
CAS 461-81-4
Molecular Formula C7H4ClF3O
Formula Weight 196.56
MDL Number MFCD00040953
Synonym 4-(Trifluoromethoxy)chlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2143709

  SDS
alfa aesar™
B2143709
10g Each for $171.77
Description & Specifications

Specifications

Formula Weight 196.56
Chemical Name or Material 1-Chloro-4-(trifluoromethoxy)benzene
Assay 98%
Molecular Formula C7H4ClF3O
MDL Number MFCD00040953
CAS 461-81-4
Synonym 4-(Trifluoromethoxy)chlorobenzene
Boiling Point 142° to 143°C
Quantity 10g