Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-4-nonyne, 94%

$52.96 - $169.32

Chemical Identifiers

Chemical Name or Material 1-Chloro-4-nonyne
CAS 3416-74-8
Molecular Formula C9H15Cl
Linear Formula CH3(CH2)3C≡C(CH2)3Cl
Formula Weight 158.67
MDL Number MFCD00041554
Beilstein 1699510
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1049103

 
alfa aesar™
L1049103
1g Each for $52.96

AAL1049106

 
alfa aesar™
L1049106
5g Each for $169.32
Description & Specifications

Specifications

Formula Weight 158.67
Chemical Name or Material 1-Chloro-4-nonyne
Assay 94%
Molecular Formula C9H15Cl
CAS 3416-74-8
Linear Formula CH3(CH2)3C≡C(CH2)3Cl
MDL Number MFCD00041554
Beilstein 1699510
Odor Odorless
Boiling Point 116° to 117°C (40mmHg)
Quantity 1g