missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-4-nonyne, 94%

Manufacturer:  Alfa Aesar™ L1049103

 View more versions of this product

Catalog No. AAL1049103



Specifications

Specifications

1-Chloro-4-nonyne
116°C to 117°C (40mmHg)
3416-74-8
CH3(CH2)3C≡C(CH2)3Cl
1699510
RHZYERSKMQJUCP-UHFFFAOYSA-N
1-chloronon-4-yne
137925
94%
Odorless
1g
C9H15Cl
MFCD00041554
1-chloro-4-nonyne, acmc-20aovu, 1-chloro-non-4-yne, 1-chloranylnon-4-yne, 4-nonyne, 1-chloro
CCCCC#CCCCCl
158.669
158.67

Chemical Identifiers

3416-74-8
158.669
RHZYERSKMQJUCP-UHFFFAOYSA-N
137925
CCCCC#CCCCCl
C9H15Cl
MFCD00041554
1-chloro-4-nonyne, acmc-20aovu, 1-chloro-non-4-yne, 1-chloranylnon-4-yne, 4-nonyne, 1-chloro
1-chloronon-4-yne
Documents