Learn More
1-Chloro-4-nitrobenzene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A153960B
Description
1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-4-nitrobenzene is an important intermediate used in the preparation of 2,4-dinitrochlorobenzene and 3,4-dichloronitrobenzene. It is used as a raw material in the preparation of 4-nitrodiphenylamine, 4-chloroaninline and (4-[(4-aminobenzene)sulfonyl]aniline). It serves as a common antioxidant, which is found in rubber.
Solubility
Soluble in boiling hot ethanol, ether, toluene, acetone, organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
1-Chloro-4-nitrobenzene | |
82°C to 86°C | |
238°C to 242°C | |
C6H4ClNO2 | |
1000 g | |
508691 | |
4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene | |
CZGCEKJOLUNIFY-UHFFFAOYSA-N | |
1-chloro-4-nitrobenzene | |
7474 | |
157.56 |
100-00-5 | |
1.515 | |
127°C (260°F) | |
MFCD00007285 | |
UN1578 | |
14,2151 | |
Soluble in boiling hot ethanol,ether,toluene,acetone,organic solvents and carbon disulfide. Sparingly soluble in cold alcohol. Insoluble in water. | |
C1=CC(=CC=C1[N+](=O)[O-])Cl | |
157.553 | |
CHEBI:34399 | |
≥98% |
Chemical Identifiers
100-00-5 | |
157.553 | |
CZGCEKJOLUNIFY-UHFFFAOYSA-N | |
7474 | |
1-chloro-4-nitrobenzene |
C6H4ClNO2 | |
MFCD00007285 | |
4-chloronitrobenzene, p-chloronitrobenzene, p-nitrochlorobenzene, benzene, 1-chloro-4-nitro, 4-nitrochlorobenzene, pncb, 4-chloro-1-nitrobenzene, p-nitrophenyl chloride, p-nitrochlorobenzol, p-nitroclorobenzene | |
CHEBI:34399 | |
C1=CC(=CC=C1[N+](=O)[O-])Cl |
Safety and Handling
GHS H Statement
H301-H311-H331-H341-H351-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P312-P330-P361-P363-P501c
H301+H311+H331-H341-H351-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID
EINECSNumber : 202-809-6
RTECSNumber : CZ1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only