missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-4-fluorobenzene, 98%, ACROS Organics™

Manufacturer:  Acros Organics 208200250

 View more versions of this product

Catalog No. AC208200250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Chloro-4-fluorobenzene
Glass bottle
Colorless to Yellow
98%
97.5
98%
ClC6H4F
05, 201
RJCGZNCCVKIBHO-UHFFFAOYSA-N
1-chloro-4-fluorobenzene
9604
Liquid
Authentic
1.4940 to 1.4960 (20°C, 589nm)
25mL
352-33-0
100.0
C6H4ClF
MFCD00000603
p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
C1=CC(=CC=C1F)Cl
130.546
130.55
≥97.5%

Chemical Identifiers

352-33-0
130.546
RJCGZNCCVKIBHO-UHFFFAOYSA-N
9604
C1=CC(=CC=C1F)Cl
C6H4ClF
MFCD00000603
p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
1-chloro-4-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-521-1

TSCA : TSCA

SDS Documents