Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-4-fluoro-2-nitrobenzene, 98+%

Chemical Identifiers

Chemical Name or Material 1-Chloro-4-fluoro-2-nitrobenzene
CAS 345-17-5
Molecular Formula C6H3ClFNO2
Formula Weight 175.55
MDL Number MFCD00084853
UN Number UN2811
Beilstein 2094191
Synonym 2-Chloro-5-fluoronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1140006

  SDS
alfa aesar™
A1140006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1140003

  SDS
alfa aesar™
A1140003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1140014

  SDS
alfa aesar™
A1140014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.55
Odor Characteristic
Molecular Formula C6H3ClFNO2
CAS 345-17-5
Health Hazard 2 GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 237° to 239°C
Chemical Name or Material 1-Chloro-4-fluoro-2-nitrobenzene
Assay ≥98%
MDL Number MFCD00084853
UN Number UN2811
Beilstein 2094191
Synonym 2-Chloro-5-fluoronitrobenzene
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Flash Point >110°C (230°F)
Melting Point 37° to 39°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.