missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Chloro-3-(trifluoromethoxy)benzene, 98%, Thermo Scientific™
$161.07 - $511.84
Chemical Identifiers
CAS | 772-49-6 |
---|---|
Molecular Formula | C7H4ClF3O |
Molecular Weight (g/mol) | 196.55 |
MDL Number | MFCD00276971 |
InChI Key | OLDJEKBXICPMAS-UHFFFAOYSA-N |
Synonym | 1-chloro-3-trifluoromethoxy benzene, 3-trifluoromethoxy chlorobenzene, m-chlorotrifluoromethoxybenzene, benzene, 1-chloro-3-trifluoromethoxy, 3-chloro-alpha,alpha,alpha-trifluoroanisole, pubchem19754, acmc-209p8q, ksc495a8t, 3-trifluoromethoxychlorobenzene |
PubChem CID | 2777270 |
IUPAC Name | 1-chloro-3-(trifluoromethoxy)benzene |
SMILES | FC(F)(F)OC1=CC(Cl)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6180514
|
Thermo Scientific Chemicals
H6180514 |
25 g |
N/A
|
|
|||||
AAH6180522
|
Thermo Scientific Chemicals
H6180522 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1-Chloro-3-(trifluoromethoxy)benzene is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
772-49-6 | |
196.55 | |
OLDJEKBXICPMAS-UHFFFAOYSA-N | |
2777270 | |
FC(F)(F)OC1=CC(Cl)=CC=C1 |
C7H4ClF3O | |
MFCD00276971 | |
1-chloro-3-trifluoromethoxy benzene, 3-trifluoromethoxy chlorobenzene, m-chlorotrifluoromethoxybenzene, benzene, 1-chloro-3-trifluoromethoxy, 3-chloro-alpha,alpha,alpha-trifluoroanisole, pubchem19754, acmc-209p8q, ksc495a8t, 3-trifluoromethoxychlorobenzene | |
1-chloro-3-(trifluoromethoxy)benzene |
Specifications
772-49-6 | |
25 g | |
C7H4ClF3O | |
1-chloro-3-trifluoromethoxy benzene, 3-trifluoromethoxy chlorobenzene, m-chlorotrifluoromethoxybenzene, benzene, 1-chloro-3-trifluoromethoxy, 3-chloro-alpha,alpha,alpha-trifluoroanisole, pubchem19754, acmc-209p8q, ksc495a8t, 3-trifluoromethoxychlorobenzene | |
OLDJEKBXICPMAS-UHFFFAOYSA-N | |
1-chloro-3-(trifluoromethoxy)benzene | |
2777270 | |
98% |
140°C | |
1.436 | |
MFCD00276971 | |
Slightly soluble in water. | |
FC(F)(F)OC1=CC(Cl)=CC=C1 | |
196.55 | |
196.55 | |
1-Chloro-3-(trifluoromethoxy)benzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C