Please login to your online account to display your discounted pricing

1-Chloro-3-nitrobenzene, 98%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  203990050

 View more versions of this product

Catalog No. AC203990050

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Yellow to brownish
Physical Form Crystalline Solid
Assay ≥97.5%
Formula Weight 157.56
Quantity 5g
EINECS Number 204-496-1
TSCA TSCA
Melting Point 41° to 45°C
RTECS Number CZ0940000
Merck Index 15, 2153
Beilstein 05, 243
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula ClC6H4NO2
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material 1-Chloro-3-nitrobenzene
CAS Min % 97.5
MDL Number MFCD00007202
Molecular Formula C6H4ClNO2
CAS 121-73-3
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.

Specifications

Chemical Name or Material 1-Chloro-3-nitrobenzene
Name Note 98%
CAS 121-73-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4ClNO2
Linear Formula ClC6H4NO2
Formula Weight 157.56
MDL Number MFCD00007202
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.

SDS & Safety Documents