Please login to your online account to display your discounted pricing

1-Chloro-3-nitrobenzene, 98%, Acros Organics

$20.17 - $58.20

Chemical Identifiers

Chemical Name or Material 1-Chloro-3-nitrobenzene
Name Note 98%
CAS 121-73-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4ClNO2
Linear Formula ClC6H4NO2
Formula Weight 157.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC203990010

  SDS
thermo fisher scientific
203990010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203990050

  SDS
thermo fisher scientific
203990050
Glass bottle 5g Each for $20.17

AC203992500

  SDS
thermo fisher scientific
203992500
Each for $58.20
Description & Specifications

Specifications

Color Yellow to brownish
Physical Form Crystalline Solid
Formula Weight 157.56
Assay ≥97.5%
Quantity 1kg
Beilstein 05, 243
Merck Index 15, 2153
RTECS Number CZ0940000
TSCA TSCA
Melting Point 41° to 45°C
EINECS Number 204-496-1
CAS Max % 100.0
Assay Percent Range 98%
MDL Number MFCD00007202
Infrared Spectrum Authentic
CAS 121-73-3
Linear Formula ClC6H4NO2
CAS Min % 97.5
Chemical Name or Material 1-Chloro-3-nitrobenzene
Name Note 98%
Molecular Formula C6H4ClNO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.