Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-3-methylbutane, 98%

Chemical Identifiers

Chemical Name or Material 1-Chloro-3-methylbutane
CAS 107-84-6
Molecular Formula C5H11Cl
Formula Weight 106.6
MDL Number MFCD00013693
UN Number UN1107
Beilstein 1730988
Merck Index 14,5116
Synonym Isoamyl chloride; Isopentyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1137109

  SDS
alfa aesar™
L1137109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1137118

  SDS
alfa aesar™
L1137118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 106.6
CAS 107-84-6
Chemical Name or Material 1-Chloro-3-methylbutane
Synonym Isoamyl chloride; Isopentyl chloride
Assay 98%
TSCA TSCA
Flash Point 16°C (60°F)
MDL Number MFCD00013693
Melting Point −104°C
Quantity 10g
Health Hazard 1 Danger
Merck Index 14,5116
Beilstein 1730988
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES
Molecular Formula C5H11Cl
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 99° to 100°C
UN Number UN1107
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.