missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-3-iodobenzene, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 221910250

 View more versions of this product

Catalog No. AC221910250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
238.45
Clear light yellow
100.0
Authentic
C6H4ClI
625-99-0
97.5
1-chloro-3-iodobenzene
Liquid
25g
05, 220
Glass bottle
ClC6H4I
98%
MFCD00001046
1.6300 to 1.6320 (20°C, 589nm)

Chemical Identifiers

625-99-0
100.0
C6H4ClI
238.45
1-chloro-3-iodobenzene
97.5
98%
ClC6H4I
MFCD00001046
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.