Learn More
1-Chloro-3-hydroxypropane, 98%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 109295000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Chloro-3-hydroxypropane | |
627-30-5 | |
100.0 | |
160°C to 162°C | |
Authentic | |
Glass bottle | |
1.4440 to 1.446 | |
MFCD00002943 | |
01, 356 | |
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1 | |
LAMUXTNQCICZQX-UHFFFAOYSA-N | |
3-chloropropan-1-ol | |
0.1% Na2CO3 | |
6 mPa.s (20°C) | |
98% |
Stabilized | |
97.5 | |
1.1300g/mL | |
75°C | |
97.5% min. (GC) | |
C3H7ClO | |
Cl(CH2)3OH | |
500g | |
1.13 | |
Solubility in water: 300g/L water (20°C). Other solubilities: reacts with water forming hcl | |
C(CO)CCl | |
94.54 | |
12313 | |
94.54 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 210-992-9
Recommended Storage : Normal conditions
RUO â Research Use Only