Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-3-fluorobenzene, 99%

Chemical Identifiers

Chemical Name or Material 1-Chloro-3-fluorobenzene
CAS 625-98-9
Molecular Formula C6H4ClF
Formula Weight 130.55
MDL Number MFCD00000569
UN Number UN1993
Beilstein 2039303
Synonym 3-Chlorofluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1312314

  SDS
alfa aesar™
A1312314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1312322

  SDS
alfa aesar™
A1312322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1312336

  SDS
alfa aesar™
A1312336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.55
Health Hazard 2 GHS H Statement
H225-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Danger
Boiling Point 126° to 128°C
Molecular Formula C6H4ClF
MDL Number MFCD00000569
Synonym 3-Chlorofluorobenzene
Assay 99%
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
TSCA TSCA
UN Number UN1993
Beilstein 2039303
Flash Point 20°C (68°F)
Chemical Name or Material 1-Chloro-3-fluorobenzene
CAS 625-98-9

Applications
1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.