Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-3-fluoro-2-propanol, 95%

Chemical Identifiers

Chemical Name or Material 1-Chloro-3-fluoro-2-propanol
CAS 453-11-2
Molecular Formula C3H6ClFO
Formula Weight 112.53
MDL Number MFCD00045284
UN Number UN1992
Beilstein 1732501
Synonym 1-Chloro-3-fluoroisopropanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1728606

 
alfa aesar™
L1728606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.53
CAS 453-11-2
Chemical Name or Material 1-Chloro-3-fluoro-2-propanol
Assay 95%
Molecular Formula C3H6ClFO
MDL Number MFCD00045284
Beilstein 1732501
Synonym 1-Chloro-3-fluoroisopropanol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 153° to 156°C
Quantity 5g
UN Number UN1992
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Applications
It is a pharmaceutical intermediate.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.