Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-3,5-difluorobenzene, 97%

Chemical Identifiers

Chemical Name or Material 1-Chloro-3,5-difluorobenzene
CAS 1435-43-4
Molecular Formula C6H3ClF2
Formula Weight 148.54
MDL Number MFCD00041518
UN Number UN1993
Beilstein 2205670
Synonym 3,5-Difluorochlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1077203

  SDS
alfa aesar™
L1077203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1077206

  SDS
alfa aesar™
L1077206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.54
Chemical Name or Material 1-Chloro-3,5-difluorobenzene
CAS 1435-43-4
Health Hazard 1 Warning
MDL Number MFCD00041518
UN Number UN1993
Synonym 3,5-Difluorochlorobenzene
Quantity 1g
Beilstein 2205670
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 29°C (84°F)
Odor Aromatic
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Assay 97%
Boiling Point 117° to 119°C
Molecular Formula C6H3ClF2
Safety and Handling

Warning

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.