missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-3,5-difluorobenzene, 97%

Manufacturer:  Alfa Aesar™ L1077203

 View more versions of this product

Catalog No. AAL1077203



Specifications

Specifications

1-Chloro-3,5-difluorobenzene
Aromatic
1g
C6H3ClF2
UN1993
3,5-difluorochlorobenzene, benzene, 1-chloro-3,5-difluoro, benzene,1-chloro-3,5-difluoro, 1-chloro-3,5-difluoro-benzene, 3,5-difluorochlorobenzene, 5-difluorochlorobenzene, pubchem4318, 3,5-difluorchlorbenzol, acmc-209cqr, intermediates-zcf02092, 1-chloro-3,5difluorobenzene
C1=C(C=C(C=C1F)Cl)F
148.537
148.54
29°C (84°F)
117°C to 119°C
1435-43-4
MFCD00041518
2205670
RFKBODCWHNDUTJ-UHFFFAOYSA-N
1-chloro-3,5-difluorobenzene
137000
97%

Chemical Identifiers

1435-43-4
148.537
RFKBODCWHNDUTJ-UHFFFAOYSA-N
137000
C1=C(C=C(C=C1F)Cl)F
C6H3ClF2
MFCD00041518
3,5-difluorochlorobenzene, benzene, 1-chloro-3,5-difluoro, benzene,1-chloro-3,5-difluoro, 1-chloro-3,5-difluoro-benzene, 3,5-difluorochlorobenzene, 5-difluorochlorobenzene, pubchem4318, 3,5-difluorchlorbenzol, acmc-209cqr, intermediates-zcf02092, 1-chloro-3,5difluorobenzene
1-chloro-3,5-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents