Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-3,3-dimethylbutane, 97%

Chemical Identifiers

Chemical Name or Material 1-Chloro-3,3-dimethylbutane
CAS 2855-08-5
Molecular Formula C6H13Cl
Linear Formula (CH3)3CCH2CH2Cl
Formula Weight 120.62
MDL Number MFCD00018980
UN Number UN1993
Beilstein 1731465
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1175309

 
alfa aesar™
L1175309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1175318

 
alfa aesar™
L1175318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 120.62
Molecular Formula C6H13Cl
Linear Formula (CH3)3CCH2CH2Cl
MDL Number MFCD00018980
CAS 2855-08-5
UN Number UN1993
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Chloro-3,3-dimethylbutane
Assay 97%
Beilstein 1731465
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 7°C (44°F)
Boiling Point 116° to 118°C
Quantity 10g

Applications
1-Chloro-3,3-dimethylbutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.