missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2-octyne, 98%

Manufacturer:  Alfa Aesar™ H5345214

 View more versions of this product

Catalog No. AAH5345214



Specifications

Specifications

1-Chloro-2-octyne
68°C (154°F)
81°C (15mmHg)
51575-83-8
CH3(CH2)4C≡CCH2Cl
1-chloro-2-octyne
CCCCCC#CCCl
144.642
144.64
0.931
1.46
25g
C8H13Cl
MFCD00274223
OUMUOQQKEAGFCJ-UHFFFAOYSA-N
1-chlorooct-2-yne
4349444
98%

Chemical Identifiers

51575-83-8
144.642
OUMUOQQKEAGFCJ-UHFFFAOYSA-N
4349444
CCCCCC#CCCl
C8H13Cl
MFCD00274223
1-chloro-2-octyne
1-chlorooct-2-yne
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents