missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2-nitrobenzene 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 109621000

 View more versions of this product

Catalog No. AC109621000


 

Specifications Description & Specifications

Specifications

1-Chloro-2-nitrobenzene
88-73-3
Authentic
Glass bottle
100mL
Fused Crystalline Mass
99+%
ClC6H4NO2
MFCD00007061
15, 2153
99+%
99.0
100.0
32° to 34°C
Yellow
≥99.0%
C6H4ClNO2
157.56
05, 241

Chemical Identifiers

88-73-3
100.0
C6H4ClNO2
157.56
1-Chloro-2-nitrobenzene, 99+%
99.0
99+%
ClC6H4NO2
MFCD00007061
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Toxic in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-854-9

RTECSNumber : CZ0875000

TSCA : TSCA