missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2-nitrobenzene, 99%

Manufacturer:  Alfa Aesar™ A1540330

 View more versions of this product

Catalog No. AAA1540330



Description

Description

Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

Solubility
Soluble in water 0.59 g/L.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

1-Chloro-2-nitrobenzene
245°C to 246°C
250g
C6H4ClNO2
UN1578
14,2151
BFCFYVKQTRLZHA-UHFFFAOYSA-N
1-chloro-2-nitrobenzene
6945
157.56
126°C (258°F)
31°C to 34°C
88-73-3
MFCD00007061
509273
2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])Cl
157.553
CHEBI:34270
99%

Chemical Identifiers

88-73-3
157.553
BFCFYVKQTRLZHA-UHFFFAOYSA-N
6945
1-chloro-2-nitrobenzene
C6H4ClNO2
MFCD00007061
2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
CHEBI:34270
C1=CC=C(C(=C1)[N+](=O)[O-])Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H351-H361-H373-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID

RTECSNumber : CZ0875000

TSCA : TSCA

Documents