Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2-nitrobenzene, 99%

Chemical Identifiers

Chemical Name or Material 1-Chloro-2-nitrobenzene
CAS 88-73-3
Molecular Formula C6H4ClNO2
Formula Weight 157.56
MDL Number MFCD00007061
UN Number UN1578
Beilstein 509273
Merck Index 14,2151
Synonym 2-Nitrochlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA154030B

 
alfa aesar™
A154030B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA154030I

 
alfa aesar™
A154030I
5000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1540330

 
alfa aesar™
A1540330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.56
Chemical Name or Material 1-Chloro-2-nitrobenzene
TSCA TSCA
Melting Point 31° to 34°C
Boiling Point 245° to 246°C
Quantity 1000g
Flash Point 126°C (258°F)
Assay 99%
Molecular Formula C6H4ClNO2
MDL Number MFCD00007061
CAS 88-73-3
UN Number UN1578
Beilstein 509273
Merck Index 14,2151
Synonym 2-Nitrochlorobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H351-H361-H373-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID
RTECS Number CZ0875000

Applications
1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives. It is important as a precursor to other compounds due to the two reactive sites present on the molecule.

Solubility
Soluble in water 0.59 g/L.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H351-H361-H373-H317
Toxic if swallowed.
Toxic in contact with skin.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.