missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Chloro-2-methyl-2-phenylpropane, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC124340010
Specifications
1-Chloro-2-methyl-2-phenylpropane | |
96°C | |
Glass bottle | |
1.04 | |
Colorless to Yellow | |
98% | |
97.5 | |
97.5% min. (GC) | |
C6H5C(CH3)2CH2Cl | |
05,II,32 | |
1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, 2-chloro-1,1-dimethylethyl benzene, 2-chloromethyl-2-phenylpropane, 2-methyl-2-phenylpropyl chloride, benzene, 2-chloro-1,1-dimethylethyl, beta-chloro-tert-butylbenzene, neophyl chloride, neophylchloride, unii-of6e0410nf | |
DNXXUUPUQXSUFH-UHFFFAOYSA-N | |
(1-chloro-2-methylpropan-2-yl)benzene | |
68191 | |
Liquid |
1.0400g/mL | |
Authentic | |
1.5230 to 1.525 | |
95°C to 100°C (10.0mmHg) | |
1L | |
515-40-2 | |
100.0 | |
C10H13Cl | |
MFCD00000940 | |
15,6544 | |
Solubility in water: insoluble | |
CC(C)(CCl)C1=CC=CC=C1 | |
168.67 | |
168.67 | |
98% |
Chemical Identifiers
515-40-2 | |
168.67 | |
DNXXUUPUQXSUFH-UHFFFAOYSA-N | |
68191 | |
CC(C)(CCl)C1=CC=CC=C1 |
C10H13Cl | |
MFCD00000940 | |
1-chloro-2-methyl-2-phenylpropane, 1-chloro-2-methylpropan-2-yl benzene, 2-chloro-1,1-dimethylethyl benzene, 2-chloromethyl-2-phenylpropane, 2-methyl-2-phenylpropyl chloride, benzene, 2-chloro-1,1-dimethylethyl, beta-chloro-tert-butylbenzene, neophyl chloride, neophylchloride, unii-of6e0410nf | |
(1-chloro-2-methylpropan-2-yl)benzene |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-197-7
TSCA : TSCA