Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2-iodotetrafluoroethane, 97%

Chemical Identifiers

Chemical Name or Material 1-Chloro-2-iodotetrafluoroethane
CAS 421-78-3
Molecular Formula ICF2CClF2
Formula Weight 262.37
MDL Number MFCD00069092
UN Number UN2810
Beilstein 1699458
Synonym 2-Chloro-1,1,2,2-tetrafluoroethyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1643006

  SDS
alfa aesar™
L1643006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1643014

  SDS
alfa aesar™
L1643014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.37
Chemical Name or Material 1-Chloro-2-iodotetrafluoroethane
CAS 421-78-3
Health Hazard 1 Danger
MDL Number MFCD00069092
Synonym 2-Chloro-1,1,2,2-tetrafluoroethyl iodide
Assay 97%
Quantity 5g
Molecular Formula ICF2CClF2
Health Hazard 2 GHS H Statement
H331-H351-H302-H312
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2810
Beilstein 1699458
Boiling Point 56°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H351-H302-H312
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.

2° to 8°C