Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2-cycloheptylethane, 96%

$299.85 - $299.85

Chemical Identifiers

Chemical Name or Material 1-Chloro-2-cycloheptylethane
Molecular Formula C9H17Cl
Formula Weight 160.69
MDL Number MFCD01632144
Synonym 2-Cycloheptylethyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2160414

  SDS
alfa aesar™
B2160414
25g Each for $299.85
Description & Specifications

Specifications

Formula Weight 160.69
Chemical Name or Material 1-Chloro-2-cycloheptylethane
MDL Number MFCD01632144
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2-Cycloheptylethyl chloride
Quantity 25g
Assay 96%
Health Hazard 1 Warning
Molecular Formula C9H17Cl
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.