missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2,5-dibromobenzene, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 263410050

 View more versions of this product

Catalog No. AC263410050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Chloro-2, 5-dibromobenzene
Glass bottle
39°C to 42°C
99+%
99.0
99+%
MFCD00018094
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
C1=CC(=C(C=C1Br)Cl)Br
270.34
270.34
99+%
Authentic
White
5g
3460-24-0
100.0
C6H3ClBr2
1,4-bis bromanyl-2-chloranyl-benzene, 1-chloro-2,5-dibromobenzene, 2,5-dibromochlorobenzene, 2-chloro-1,4-dibromobenzene, acmc-209i8v, benzene, 1,4-dibromo-2-chloro, chloro-3,6-dibrom obenzene, pubchem3633
LOWQAATYMJIWOG-UHFFFAOYSA-N
1,4-dibromo-2-chlorobenzene
2724603
Fine Crystalline Powder

Chemical Identifiers

3460-24-0
270.34
LOWQAATYMJIWOG-UHFFFAOYSA-N
2724603
C1=CC(=C(C=C1Br)Cl)Br
C6H3ClBr2
MFCD00018094
1,4-bis bromanyl-2-chloranyl-benzene, 1-chloro-2,5-dibromobenzene, 2,5-dibromochlorobenzene, 2-chloro-1,4-dibromobenzene, acmc-209i8v, benzene, 1,4-dibromo-2-chloro, chloro-3,6-dibrom obenzene, pubchem3633
1,4-dibromo-2-chlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents