missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Chloro-2,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific™
$60.81 - $60.81
Chemical Identifiers
CAS | 328-91-6 |
---|---|
Molecular Formula | C8H3ClF6 |
Molecular Weight (g/mol) | 248.552 |
MDL Number | MFCD00632388 |
InChI Key | IDNVTCBYGYOPKJ-UHFFFAOYSA-N |
Synonym | 2,5-bis trifluoromethyl chlorobenzene, 2-chloro-1,4-bis trifluoromethyl benzene, 1-chloro-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-chlorobenzene, 2,5-ditrifluoromethyl-chlorobenzene, 2-chlor-1,4-bis-trifluormethyl-benzol, 2-chloro-1,4-bis trifluoromethyl benzene #, benzene, 1-chloro-2,5-bis trifluoromethyl, benzene, 2-chloro-1,4-bis trifluoromethyl |
PubChem CID | 619189 |
IUPAC Name | 2-chloro-1,4-bis(trifluoromethyl)benzene |
SMILES | C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6429703
|
Thermo Scientific Chemicals
H6429703 |
1 g |
Each for $60.81
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt can be used as a synthetic building block.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
328-91-6 | |
248.552 | |
IDNVTCBYGYOPKJ-UHFFFAOYSA-N | |
619189 | |
C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F |
C8H3ClF6 | |
MFCD00632388 | |
2,5-bis trifluoromethyl chlorobenzene, 2-chloro-1,4-bis trifluoromethyl benzene, 1-chloro-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-chlorobenzene, 2,5-ditrifluoromethyl-chlorobenzene, 2-chlor-1,4-bis-trifluormethyl-benzol, 2-chloro-1,4-bis trifluoromethyl benzene #, benzene, 1-chloro-2,5-bis trifluoromethyl, benzene, 2-chloro-1,4-bis trifluoromethyl | |
2-chloro-1,4-bis(trifluoromethyl)benzene |
Specifications
328-91-6 | |
147°C to 149°C | |
1.4135 | |
1 g | |
2,5-bis trifluoromethyl chlorobenzene, 2-chloro-1,4-bis trifluoromethyl benzene, 1-chloro-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-chlorobenzene, 2,5-ditrifluoromethyl-chlorobenzene, 2-chlor-1,4-bis-trifluormethyl-benzol, 2-chloro-1,4-bis trifluoromethyl benzene #, benzene, 1-chloro-2,5-bis trifluoromethyl, benzene, 2-chloro-1,4-bis trifluoromethyl | |
IDNVTCBYGYOPKJ-UHFFFAOYSA-N | |
2-chloro-1,4-bis(trifluoromethyl)benzene | |
619189 | |
98% | |
1-Chloro-2,5-bis(trifluoromethyl)benzene |
1.504 g/mL | |
C8H3ClF6 | |
MFCD00632388 | |
1978018 | |
Slightly soluble in water. | |
C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F | |
248.552 | |
248.55 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures