missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Chloro-2,4-dinitrobenzene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 160515000

 View more versions of this product

Catalog No. AC160515000



Specifications

Specifications

1-Chloro-2, 4-dinitrobenzene
Authentic
315°C
48°C to 53°C
99%
98.5
98.5% min. (GC)
ClC6H3(NO2)2
05, 263
1,3-dinitro-4-chlorobenzene
VYZAHLCBVHPDDF-UHFFFAOYSA-N
1-chloro-2,4-dinitrobenzene
6
202.55
99%
186°C
Plastic bottle
Yellow to Brown
500g
97-00-7
100.0
C6H3ClN2O4
MFCD00007075
15, 2137
Solubility in water: insoluble. Other solubilities: soluble in hot alcohol,ether,benzene and,carbon disulfide
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Cl
202.55
CHEBI:34718
Crystalline Mass, Chunks or Crystals and Powder

Chemical Identifiers

97-00-7
202.55
VYZAHLCBVHPDDF-UHFFFAOYSA-N
6
1-chloro-2,4-dinitrobenzene
C6H3ClN2O4
MFCD00007075
1,3-dinitro-4-chlorobenzene
CHEBI:34718
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 202-551-4

RTECSNumber : CZ0525000

TSCA : TSCA

SDS Documents