missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2,4-difluorobenzene, 98%

Chemical Identifiers

CAS 1435-44-5
Molecular Formula C6H3ClF2
Formula Weight 148.54
MDL Number MFCD00042569
UN Number UN1993
Beilstein 2081077
Synonym 2,4-Difluorochlorobenzene
Chemical Name or Material 1-Chloro-2,4-difluorobenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1549509 SDS Alfa Aesar™
A1549509
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1549518 SDS Alfa Aesar™
A1549518
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1549530 SDS Alfa Aesar™
A1549530
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1435-44-5
148.54
UN1993
2,4-Difluorochlorobenzene
C6H3ClF2
MFCD00042569
2081077
1-Chloro-2,4-difluorobenzene

Specifications

32°C (89°F)
−26°C
1435-44-5
C6H3ClF2
MFCD00042569
2081077
1-Chloro-2,4-difluorobenzene
126° to 127°C
10g
Aromatic
148.54
UN1993
2,4-Difluorochlorobenzene
98%

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.