missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2,4-difluorobenzene, 98%

Manufacturer:  Alfa Aesar™ A1549518

 View more versions of this product

Catalog No. AAA1549518



Specifications

Specifications

1-Chloro-2,4-difluorobenzene
Aromatic
-26°C
1435-44-5
MFCD00042569
2081077
AJCSNHQKXUSMMY-UHFFFAOYSA-N
1-chloro-2,4-difluorobenzene
137001
98%
32°C (89°F)
126°C to 127°C
50g
C6H3ClF2
UN1993
2,4-difluorochlorobenzene, 1-chloro-2,4-difluoro-benzene, benzene,1-chloro-2,4-difluoro, benzene, 1-chloro-2,4-difluoro, pubchem3415, 2,4-difluorchlorbenzol, 2,4-diflurochlorobenzene, acmc-1c2st, intermediates-zcf02093, 2,4-di-fluorochlorobenzene
C1=CC(=C(C=C1F)F)Cl
148.537
148.54

Chemical Identifiers

1435-44-5
148.537
AJCSNHQKXUSMMY-UHFFFAOYSA-N
137001
C1=CC(=C(C=C1F)F)Cl
C6H3ClF2
MFCD00042569
2,4-difluorochlorobenzene, 1-chloro-2,4-difluoro-benzene, benzene,1-chloro-2,4-difluoro, benzene, 1-chloro-2,4-difluoro, pubchem3415, 2,4-difluorchlorbenzol, 2,4-diflurochlorobenzene, acmc-1c2st, intermediates-zcf02093, 2,4-di-fluorochlorobenzene
1-chloro-2,4-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents