Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2,4-bis(trifluoromethyl)benzene, 97%

Chemical Identifiers

Chemical Name or Material 1-Chloro-2,4-bis(trifluoromethyl)benzene
CAS 327-76-4
Molecular Formula C8H3ClF6
Formula Weight 248.56
MDL Number MFCD01631638
UN Number UN1993
Beilstein 1978012
Synonym 2,4-Bis(trifluoromethyl)chlorobenzene; 4-Chloro-1,3-bis(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2273403

  SDS
alfa aesar™
B2273403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2273406

  SDS
alfa aesar™
B2273406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.56
Chemical Name or Material 1-Chloro-2,4-bis(trifluoromethyl)benzene
Assay 97%
Molecular Formula C8H3ClF6
MDL Number MFCD01631638
CAS 327-76-4
UN Number UN1993
Beilstein 1978012
Synonym 2,4-Bis(trifluoromethyl)chlorobenzene; 4-Chloro-1,3-bis(trifluoromethyl)benzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 59°C (138°F)
Boiling Point 150° to 154°C
Melting Point −59°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.