Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2,3,4-trifluorobenzene, 99+%

$51.11 - $571.07

Chemical Identifiers

Chemical Name or Material 1-Chloro-2,3,4-trifluorobenzene
CAS 36556-42-0
Molecular Formula C6H2ClF3
Formula Weight 166.53
MDL Number MFCD00042222
Synonym 2,3,4-Trifluorochlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2306006

  SDS
alfa aesar™
B2306006
5g Each for $51.11

AAB2306014

  SDS
alfa aesar™
B2306014
25g Each for $181.64

AAB2306022

  SDS
alfa aesar™
B2306022
100g Each for $571.07
Description & Specifications

Specifications

Formula Weight 166.53
Chemical Name or Material 1-Chloro-2,3,4-trifluorobenzene
Assay ≥99%
Molecular Formula C6H2ClF3
MDL Number MFCD00042222
CAS 36556-42-0
Synonym 2,3,4-Trifluorochlorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.