missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%

Manufacturer:  Alfa Aesar™ L1731514

 View more versions of this product

Catalog No. AAL1731514



Specifications

Specifications

1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
Ether-like
48°C to 49°C
25g
C3H2ClF5O
1852087
1-chloro-2,2,2-trifluoroethyl difluoromethyl ether
C(C(F)(F)F)(OC(F)F)Cl
184.49
CHEBI:6015
97%
1.496g/mL
<1.3
Colorless
26675-46-7
MFCD00066609
14,5175
PIWKPBJCKXDKJR-UHFFFAOYSA-N
2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
3763
184.49

Chemical Identifiers

26675-46-7
184.49
PIWKPBJCKXDKJR-UHFFFAOYSA-N
3763
2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
C3H2ClF5O
MFCD00066609
1-chloro-2,2,2-trifluoroethyl difluoromethyl ether
CHEBI:6015
C(C(F)(F)F)(OC(F)F)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H336
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : KN6799000

Documents