missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate, 97%

$104.86 - $428.70

Chemical Identifiers

CAS 115007-14-2
Molecular Formula C9H16BClF4N2
Molecular Weight (g/mol) 274.495
MDL Number MFCD00191334
InChI Key NRROZYAIGHAMHH-UHFFFAOYSA-N
Synonym 1-chloro-1-pyrrolidinylmethylene pyrrolidinium tetrafluoroborate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium tetrafluoroborate, 1-chloro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium tetrafluoroborate, c9h16cln2.bf4, ksc173s7b, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium te, chloro-n,n,n',n'-bis tetramethylene formamidinium tetrafluoroborate, 1-chloranyl pyrrolidin-1-ium-1-ylidene methyl pyrrolidine tetrafluoroborate, 1-chloro 1-pyrrolidin-1-iumylidene methyl pyrrolidine tetrafluoroborate, 1-chloro pyrrolidin-1-ium-1-ylidene methyl pyrrolidine tetrafluoroborate
PubChem CID 2736654
IUPAC Name 1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1CCN(C1)C(=[N+]2CCCC2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6021318 View Documents Alfa Aesar™
H6021318
50g Each for $104.86
 
AAH6021330 View Documents Alfa Aesar™
H6021330
250g Each for $428.70
 
Specifications

Chemical Identifiers

115007-14-2
274.495
NRROZYAIGHAMHH-UHFFFAOYSA-N
2736654
[B-](F)(F)(F)F.C1CCN(C1)C(=[N+]2CCCC2)Cl
C9H16BClF4N2
MFCD00191334
1-chloro-1-pyrrolidinylmethylene pyrrolidinium tetrafluoroborate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium tetrafluoroborate, 1-chloro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium tetrafluoroborate, c9h16cln2.bf4, ksc173s7b, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium te, chloro-n,n,n',n'-bis tetramethylene formamidinium tetrafluoroborate, 1-chloranyl pyrrolidin-1-ium-1-ylidene methyl pyrrolidine tetrafluoroborate, 1-chloro 1-pyrrolidin-1-iumylidene methyl pyrrolidine tetrafluoroborate, 1-chloro pyrrolidin-1-ium-1-ylidene methyl pyrrolidine tetrafluoroborate
1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;tetrafluoroborate

Specifications

50g
C9H16BClF4N2
1-chloro-1-pyrrolidinylmethylene pyrrolidinium tetrafluoroborate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium tetrafluoroborate, 1-chloro pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium tetrafluoroborate, c9h16cln2.bf4, ksc173s7b, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium te, chloro-n,n,n',n'-bis tetramethylene formamidinium tetrafluoroborate, 1-chloranyl pyrrolidin-1-ium-1-ylidene methyl pyrrolidine tetrafluoroborate, 1-chloro 1-pyrrolidin-1-iumylidene methyl pyrrolidine tetrafluoroborate, 1-chloro pyrrolidin-1-ium-1-ylidene methyl pyrrolidine tetrafluoroborate
[B-](F)(F)(F)F.C1CCN(C1)C(=[N+]2CCCC2)Cl
274.495
274.49
1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium tetrafluoroborate
115007-14-2
MFCD00191334
NRROZYAIGHAMHH-UHFFFAOYSA-N
1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;tetrafluoroborate
2736654
97%