missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate, 98+%

Manufacturer:  Alfa Aesar™ H5424703

 View more versions of this product

Catalog No. AAH5424703



Specifications

Specifications

1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
1g
C9H16ClF6N2P
7898575
NHEGCUSBUWGOQM-UHFFFAOYSA-N
1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
2736652
≥98%
153°C to 156°C
135540-11-3
MFCD00191333
1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v
C1CCN(C1)C(=[N+]2CCCC2)Cl.F[P-](F)(F)(F)(F)F
332.655
332.65

Chemical Identifiers

135540-11-3
332.655
NHEGCUSBUWGOQM-UHFFFAOYSA-N
2736652
C1CCN(C1)C(=[N+]2CCCC2)Cl.F[P-](F)(F)(F)(F)F
C9H16ClF6N2P
MFCD00191333
1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v
1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents