missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-1,2,2-trifluorocyclobutane, 97%

$55.51 - $175.80

Chemical Identifiers

CAS 661-71-2
Molecular Formula C4H4ClF3
Molecular Weight (g/mol) 144.521
MDL Number MFCD00042139
InChI Key ZZQYDYODFHABLC-UHFFFAOYSA-N
Synonym cyclobutane, 1-chloro-1,2,2-trifluoro, 1-ctfcb, d0p0zb, cyclobutane,1-chloro-1,2,2-trifluoro
PubChem CID 79091
IUPAC Name 1-chloro-1,2,2-trifluorocyclobutane
SMILES C1CC(C1(F)F)(F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1643606 View Documents Alfa Aesar™
L1643606
5g Each for $55.51
 
AAL1643614 View Documents Alfa Aesar™
L1643614
25g Each for $175.80
 
Specifications

Chemical Identifiers

661-71-2
144.521
ZZQYDYODFHABLC-UHFFFAOYSA-N
79091
C1CC(C1(F)F)(F)Cl
C4H4ClF3
MFCD00042139
cyclobutane, 1-chloro-1,2,2-trifluoro, 1-ctfcb, d0p0zb, cyclobutane,1-chloro-1,2,2-trifluoro
1-chloro-1,2,2-trifluorocyclobutane

Specifications

81°C
661-71-2
MFCD00042139
1902628
ZZQYDYODFHABLC-UHFFFAOYSA-N
1-chloro-1,2,2-trifluorocyclobutane
79091
97%
5g
C4H4ClF3
UN1993
cyclobutane, 1-chloro-1,2,2-trifluoro, 1-ctfcb, d0p0zb, cyclobutane,1-chloro-1,2,2-trifluoro
C1CC(C1(F)F)(F)Cl
144.521
144.52
1-Chloro-1,2,2-trifluorocyclobutane
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.