missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Chloro-1,1,2-trifluoro-2-iodoethane, 97%

$58.92 - $436.42

Chemical Identifiers

CAS 354-26-7
Molecular Formula C2HClF3I
Molecular Weight (g/mol) 244.38
MDL Number MFCD00042138
InChI Key XONGRFUEHPBOBB-UHFFFAOYSA-N
Synonym 1-chloro-2-iodo-1,1,2-trifluoroethane, acmc-1cr56, 2-chloro-1,2,2-trifluoroethyl iodide, 1,2,2-trifluoro-2-chloro-1-iodoethane, ethane,1-chloro-1,1,2-trifluoro-2-iodo, ethane, 1-chloro-1,1,2-trifluoro-2-iodo
PubChem CID 78980
IUPAC Name 1-chloro-1,1,2-trifluoro-2-iodoethane
SMILES C(C(F)(F)Cl)(F)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1672106 View Documents Alfa Aesar™
L1672106
5g Each for $58.92
 
AAL1672114 View Documents Alfa Aesar™
L1672114
25g Each for $157.30
 
AAL1672122 View Documents Alfa Aesar™
L1672122
100g Each for $436.42
 
Specifications

Chemical Identifiers

354-26-7
244.38
XONGRFUEHPBOBB-UHFFFAOYSA-N
78980
C(C(F)(F)Cl)(F)I
C2HClF3I
MFCD00042138
1-chloro-2-iodo-1,1,2-trifluoroethane, acmc-1cr56, 2-chloro-1,2,2-trifluoroethyl iodide, 1,2,2-trifluoro-2-chloro-1-iodoethane, ethane,1-chloro-1,1,2-trifluoro-2-iodo, ethane, 1-chloro-1,1,2-trifluoro-2-iodo
1-chloro-1,1,2-trifluoro-2-iodoethane

Specifications

77°C
354-26-7
ICHFCF2Cl
1737257
XONGRFUEHPBOBB-UHFFFAOYSA-N
1-chloro-1,1,2-trifluoro-2-iodoethane
78980
97%
5g
C2HClF3I
MFCD00042138
1-chloro-2-iodo-1,1,2-trifluoroethane, acmc-1cr56, 2-chloro-1,2,2-trifluoroethyl iodide, 1,2,2-trifluoro-2-chloro-1-iodoethane, ethane,1-chloro-1,1,2-trifluoro-2-iodo, ethane, 1-chloro-1,1,2-trifluoro-2-iodo
C(C(F)(F)Cl)(F)I
244.38
244.38
1-Chloro-1,1,2-trifluoro-2-iodoethane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning