missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butylpyridinium Bromide, TCI America™

$55.00 - $146.67

Chemical Identifiers

CAS 874-80-6
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.122
MDL Number MFCD00671496
InChI Key KVBQNFMTEUEOCD-UHFFFAOYSA-M
Synonym 1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
PubChem CID 2734238
IUPAC Name 1-butylpyridin-1-ium;bromide
SMILES CCCC[N+]1=CC=CC=C1.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17435G View Documents TCI America
B17435G
5g Each for $55.00
 
B174325G View Documents TCI America
B174325G
25g Each for $146.67
 
Specifications

Chemical Identifiers

874-80-6
216.122
KVBQNFMTEUEOCD-UHFFFAOYSA-M
2734238
CCCC[N+]1=CC=CC=C1.[Br-]
C9H14BrN
MFCD00671496
1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
1-butylpyridin-1-ium;bromide

Specifications

White-Yellow
5g
C9H14BrN
1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
CCCC[N+]1=CC=CC=C1.[Br-]
216.122
216.12
≥98.0% (HPLC,T)
104°C
874-80-6
MFCD00671496
KVBQNFMTEUEOCD-UHFFFAOYSA-M
1-butylpyridin-1-ium;bromide
2734238
Crystal-Powder at 20°C
1-Butylpyridinium Bromide
Safety and Handling

Safety and Handling

TSCA : No