Please login to your online account to display your discounted pricing

1-Butylpyridinium bromide, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  356710250

 View more versions of this product

Catalog No. AC356710250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Crystals
Formula Weight 216.12
Quantity 25g
Color White to beige
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
CAS 874-80-6
Molecular Formula C9H14BrN
Chemical Name or Material 1-Butylpyridinium bromide
CAS Min % 97.5
MDL Number MFCD00671496
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material 1-Butylpyridinium bromide
CAS 874-80-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H14BrN
Formula Weight 216.12
MDL Number MFCD00671496
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents