missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butylpyridinium bromide, 98%, ACROS Organics™

$185.18 - $185.18

Chemical Identifiers

CAS 874-80-6
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.12
MDL Number MFCD00671496
InChI Key KVBQNFMTEUEOCD-UHFFFAOYSA-M
Synonym 1-butylpyridin-1-ium bromide, 1-butylpyridinium bromide, 1-butylpyridiniumbromide, 1-n-butylpyridinium bromide, acmc-209qnu, butylpyridinium bromide, f72fd1bswg, n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, unii-f72fd1bswg
PubChem CID 2734238
IUPAC Name 1-butylpyridin-1-ium;bromide
SMILES CCCC[N+]1=CC=CC=C1.[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC356710250
SDS View Documents
Thermo Fisher Scientific
356710250
25g Glass bottle
Each for $185.18
 
Description

Description

Ionic liquid
Specifications

Chemical Identifiers

874-80-6
216.12
KVBQNFMTEUEOCD-UHFFFAOYSA-M
2734238
CCCC[N+]1=CC=CC=C1.[Br-]
C9H14BrN
MFCD00671496
1-butylpyridin-1-ium bromide, 1-butylpyridinium bromide, 1-butylpyridiniumbromide, 1-n-butylpyridinium bromide, acmc-209qnu, butylpyridinium bromide, f72fd1bswg, n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, unii-f72fd1bswg
1-butylpyridin-1-ium;bromide

Specifications

Authentic
White to Beige
874-80-6
100.0
C9H14BrN
1-butylpyridin-1-ium bromide, 1-butylpyridinium bromide, 1-butylpyridiniumbromide, 1-n-butylpyridinium bromide, acmc-209qnu, butylpyridinium bromide, f72fd1bswg, n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, unii-f72fd1bswg
CCCC[N+]1=CC=CC=C1.[Br-]
216.12
216.12
98%
Glass bottle
25g
97.5
97.5% min. (Argentometry)
MFCD00671496
KVBQNFMTEUEOCD-UHFFFAOYSA-M
1-butylpyridin-1-ium;bromide
2734238
Crystalline Powder or Crystals
1-Butylpyridinium bromide
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

TSCA : TSCA

SDS