missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butylpiperazine, TCI America™

$70.28 - $241.39

Chemical Identifiers

CAS 5610-49-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00039702
InChI Key YKSVXVKIYYQWBB-UHFFFAOYSA-N
PubChem CID 424322
IUPAC Name 1-butylpiperazine
SMILES CCCCN1CCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B40785ML View Documents TCI America
B40785ML
5mL Each for $70.28
 
B407825ML View Documents TCI America
B407825ML
25mL Each for $241.39
 
Specifications

Chemical Identifiers

5610-49-1
142.246
YKSVXVKIYYQWBB-UHFFFAOYSA-N
1-butylpiperazine
C8H18N2
MFCD00039702
424322
CCCCN1CCNCC1

Specifications

192°C
5mL
C8H18N2
YKSVXVKIYYQWBB-UHFFFAOYSA-N
1-butylpiperazine
424322
Clear Liquid at 20°C
1-Butylpiperazine
Yellow
5610-49-1
MFCD00039702
CCCCN1CCNCC1
142.246
142.25
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No