Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%

$73.53 - $164.24

Chemical Identifiers

Chemical Name or Material 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
CAS 85167-10-8
Molecular Formula C14H25BO2
Formula Weight 236.17
MDL Number MFCD09037499
Synonym (1S,2S,3R,5S)-(+)-2,3-Pinanediol butylboronate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2766103

  SDS
alfa aesar™
H2766103
1g Each for $164.24

AAH27661MD

  SDS
alfa aesar™
H27661MD
250mg Each for $73.53
Description & Specifications

Specifications

Formula Weight 236.17
CAS 85167-10-8
Synonym (1S,2S,3R,5S)-(+)-2,3-Pinanediol butylboronate
MDL Number MFCD09037499
Molecular Formula C14H25BO2
Optical Rotation +25° (c=1 in Chloroform)
Chemical Name or Material 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
Assay 95%
Quantity 1g