Learn More
(1-Butyl)triphenylphosphonium chloride, Thermo Scientific Chemicals
$91.44 - $247.14
Chemical Identifiers
CAS | 13371-17-0 |
---|---|
Molecular Formula | C22H24ClP |
Molecular Weight (g/mol) | 354.86 |
MDL Number | MFCD00040546 |
InChI Key | MFIUDWFSVDFDDY-UHFFFAOYSA-M |
Synonym | butyltriphenylphosphonium chloride, butyl triphenyl phosphonium chloride, phosphonium, butyltriphenyl-, chloride, 1-butyl triphenylphosphonium chloride, phosphonium, butyltriphenyl-, chloride 1:1, butyltriphenyl phosphonium chloride, butyl triphenyl phosphanium chloride, butyltriphenylphosphanium chloride, acmc-1bzbo, butyltriphenyl-phosphoniuchloride |
PubChem CID | 166797 |
IUPAC Name | butyl(triphenyl)phosphanium;chloride |
SMILES | [Cl-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2579614
|
Thermo Scientific Chemicals
H2579614 |
25 g |
Each for $91.44
|
|
|||||
AAH2579622
|
Thermo Scientific Chemicals
H2579622 |
100 g |
Each for $247.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13371-17-0 | |
354.86 | |
MFIUDWFSVDFDDY-UHFFFAOYSA-M | |
166797 | |
[Cl-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C22H24ClP | |
MFCD00040546 | |
butyltriphenylphosphonium chloride, butyl triphenyl phosphonium chloride, phosphonium, butyltriphenyl-, chloride, 1-butyl triphenylphosphonium chloride, phosphonium, butyltriphenyl-, chloride 1:1, butyltriphenyl phosphonium chloride, butyl triphenyl phosphanium chloride, butyltriphenylphosphanium chloride, acmc-1bzbo, butyltriphenyl-phosphoniuchloride | |
butyl(triphenyl)phosphanium;chloride |
Specifications
13371-17-0 | |
C22H24ClP | |
MFCD00040546 | |
UN3464 | |
butyltriphenylphosphonium chloride, butyl triphenyl phosphonium chloride, phosphonium, butyltriphenyl-, chloride, 1-butyl triphenylphosphonium chloride, phosphonium, butyltriphenyl-, chloride 1:1, butyltriphenyl phosphonium chloride, butyl triphenyl phosphanium chloride, butyltriphenylphosphanium chloride, acmc-1bzbo, butyltriphenyl-phosphoniuchloride | |
[Cl-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
354.86 | |
354.86 |
225°C to 228°C | |
CH3(CH2)3P(C6H5)3Cl | |
25 g | |
Hygroscopic | |
MFIUDWFSVDFDDY-UHFFFAOYSA-M | |
butyl(triphenyl)phosphanium;chloride | |
166797 | |
(1-Butyl)triphenylphosphonium chloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
EINECSNumber : 236-443-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only