Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Butyl)triphenylphosphonium bromide, 99%

Chemical Identifiers

Chemical Name or Material (1-Butyl)triphenylphosphonium bromide
CAS 1779-51-7
Molecular Formula CH3(CH2)3P(C6H5)3Br
Formula Weight 399.32
MDL Number MFCD00011855
UN Number UN3464
Beilstein 3580224
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1050422

  SDS
alfa aesar™
A1050422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1050436

  SDS
alfa aesar™
A1050436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1050414

  SDS
alfa aesar™
A1050414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (1-Butyl)triphenylphosphonium bromide
CAS 1779-51-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
RTECS Number TA1855200
TSCA TSCA
Assay 99%
Molecular Formula CH3(CH2)3P(C6H5)3Br
Formula Weight 399.32
MDL Number MFCD00011855
UN Number UN3464
Beilstein 3580224
Melting Point 240° to 243°C
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.