Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Butyl isocyanate, 97%


Quantity 5g

Manufacturer: alfa aesar™  L05664-06

 View more versions of this product

Catalog No. AAL0566406

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 0.890g/mL
Refractive Index 1.4060
Quantity 5g
Color Colorless
Boiling Point 112 to 116°C
Quantity 5g
Sensitivity Moisture Sensitive
Chemical Name or Material 1-Butyl isocyanate
CAS 111-36-4
Physical Form Liquid
Assay 97%
Linear Formula CH3CH2CH2CH2NCO
Formula Weight 99.13g/mol
MDL Number MFCD00002046
UN Number UN2485
Beilstein 73917
Merck Index 14,1575
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H330-H334-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: N-BUTYL ISOCYANATE
RTECS Number NQ8250000
TSCA TSCA

Specifications

Chemical Name or Material 1-Butyl isocyanate
CAS 111-36-4
Linear Formula CH3CH2CH2CH2NCO
Formula Weight 99.13g/mol
MDL Number MFCD00002046
UN Number UN2485
Beilstein 73917
Merck Index 14,1575
Accessories

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H330-H334-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

2° to 8°C