Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Butyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 1-Butyl isocyanate
CAS 111-36-4
Linear Formula CH3CH2CH2CH2NCO
Formula Weight 99.13
MDL Number MFCD00002046
UN Number UN2485
Beilstein 73917
Merck Index 14,1575
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0566406

 
alfa aesar™
L0566406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0566414

 
alfa aesar™
L0566414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 112° to 116°C
Formula Weight 99.13
Assay 97%
MDL Number MFCD00002046
Chemical Name or Material 1-Butyl isocyanate
Physical Form Liquid
Quantity 5g
Linear Formula CH3CH2CH2CH2NCO
Beilstein 73917
Health Hazard 2 GHS H Statement
H225-H330-H334-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: N-BUTYL ISOCYANATE
TSCA TSCA
Refractive Index 1.4060
Color Colorless
Sensitivity Moisture Sensitive
CAS 111-36-4
UN Number UN2485
Health Hazard 1 Danger
Density 0.890g/mL
RTECS Number NQ8250000
Merck Index 14,1575
Storage Note 1 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H330-H334-H314-H318-H302
Highly flammable liquid and vapor.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

2° to 8°C