missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butyl-4-methylpyridinium Hexafluorophosphate, TCI America™

$84.03 - $255.14

Chemical Identifiers

CAS 401788-99-6
Molecular Formula C10H16F6NP
Molecular Weight (g/mol) 295.209
InChI Key XBSZSSGZQFAUPF-UHFFFAOYSA-N
Synonym 1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296, 1-butyl-4-methylpyridinium hexafluorophosphate
PubChem CID 11254899
IUPAC Name 1-butyl-4-methylpyridin-1-ium;hexafluorophosphate
SMILES CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27015G View Documents TCI America
B27015G
5g Each for $84.03
 
B270125G View Documents TCI America
B270125G
25g Each for $255.14
 
Specifications

Chemical Identifiers

401788-99-6
295.209
1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296, 1-butyl-4-methylpyridinium hexafluorophosphate
1-butyl-4-methylpyridin-1-ium;hexafluorophosphate
C10H16F6NP
XBSZSSGZQFAUPF-UHFFFAOYSA-N
11254899
CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F

Specifications

Undesignated
5g
C10H16F6NP
XBSZSSGZQFAUPF-UHFFFAOYSA-N
1-butyl-4-methylpyridin-1-ium;hexafluorophosphate
11254899
Crystal-Powder at 20°C
1-Butyl-4-methylpyridinium Hexafluorophosphate
42°C
401788-99-6
1-butyl-4-methylpyridinium hexafluorophosphate, 4mbppf6, 1-butyl-4-methylpyridin-1-ium hexafluorophosphate, acmc-209jbi, dsstox_cid_29152, dsstox_rid_83371, dsstox_gsid_49296, 1-butyl-4-methylpyridinium hexafluorophosphate
CCCC[N+]1=CC=C(C=C1)C.F[P-](F)(F)(F)(F)F
295.209
295.21
≥97.0% (T)
Safety and Handling

Safety and Handling

TSCA : No