missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Butyl-4-methylpyridinium Bromide, TCI America™

$111.53 - $333.06

Chemical Identifiers

CAS 65350-59-6
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.149
InChI Key UWVZAZVPOZTKNM-UHFFFAOYSA-M
PubChem CID 13487610
IUPAC Name 1-butyl-4-methylpyridin-1-ium;bromide
SMILES CCCC[N+]1=CC=C(C=C1)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27005G View Documents TCI America
B27005G
5g Each for $111.53
 
B270025G View Documents TCI America
B270025G
25g Each for $333.06
 
Specifications

Chemical Identifiers

65350-59-6
230.149
13487610
CCCC[N+]1=CC=C(C=C1)C.[Br-]
C10H16BrN
UWVZAZVPOZTKNM-UHFFFAOYSA-M
1-butyl-4-methylpyridin-1-ium;bromide

Specifications

Undesignated
5g
C10H16BrN
CCCC[N+]1=CC=C(C=C1)C.[Br-]
230.149
230.15
≥98.0% (HPLC,T)
137°C
65350-59-6
UWVZAZVPOZTKNM-UHFFFAOYSA-M
1-butyl-4-methylpyridin-1-ium;bromide
13487610
Crystal-Powder at 20°C
1-Butyl-4-methylpyridinium Bromide
Safety and Handling

Safety and Handling

TSCA : No