Learn More
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 354210050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquidSpecifications
1-Butyl-3-methylimidazolium tetrafluoroborate | |
98.0 | |
-71.0°C | |
1.2077g/mL | |
98% min. (HPLC) | |
C8H15BF4N2 | |
5 g | |
15, 1584 | |
LSBXQLQATZTAPE-UHFFFAOYSA-N | |
1-butyl-3-methylimidazol-3-ium;tetrafluoroborate | |
2734178 | |
226.02 | |
Viscous Liquid |
174501-65-6 | |
100.0 | |
Orange-Yellow | |
Authentic | |
Glass bottle | |
MFCD03095449 | |
1.2077 | |
1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate | |
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1 | |
226.03 | |
118.3 mPa.s (25°C) | |
98+% |
Chemical Identifiers
174501-65-6 | |
226.03 | |
LSBXQLQATZTAPE-UHFFFAOYSA-N | |
2734178 | |
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1 |
C8H15BF4N2 | |
MFCD03095449 | |
1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate | |
1-butyl-3-methylimidazol-3-ium;tetrafluoroborate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical adv
GHS Signal Word: Danger
TSCA : TSCA
RUO – Research Use Only